missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KIN001-266

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
KIN001-266 is a potent and selective Tpl2 (Cot; MAP3K8) inhibitor (IC50 = 1.6 nM). It exhibits 6875-fold selectivity over EGFR. In vivo, it inhibits TNF-α production in a LPS-stimulated rat inflammation model. KIN001-266 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 8-Chloro-4-[(3-chloro-4-fluorophenyl)amino]-6-[[[1-(1-ethyl-4-piperidinyl)-1H-1,2,3-triazol-4-yl]methyl]amino]-3-quinolinecarbonitrile |
| CAS | 915363-56-3 |
| Quantity | 2 mg |
| Target | MAP3K8/Tpl2/COT |
| Molecular Formula | C26H25Cl2FN8 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction