Learn More
Kimtech Science™ Wipe Delicate Task White
Product Code. 17701868 Shop All Kimberly-Clark Professional Products
Product Code. 17701868 Supplier Kimberly Clark Supplier No. 7557
In Stock
Add to basket
Edge
Add to basket
Wipe liquids with multi-ply absorbency and low lint content for intricate dusting. Kimberly Clark™ Kimtech Scientific™ Delicate Task Wipers are ideal for cleaning delicate glassware, instruments, and surface/parts.
A range of high-performance wipers, suitable for ISO Class 4 or higher laboratory environments.
Ideal for: cleaning of delicate glassware, instruments and surface/parts; wiping liquids, with multi-ply absorbency and low lint content for intricate dusting.
Available as: 100 or 200 format, white paper wipers, dispensed from a POP-UP Box flexible system, giving controlled access to hygienically protected wipers when you’re cleaning or wiping on the move.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wipe
Length (Metric) 20 cm
Width (Metric) 21 cm
Material Paper
Color White
For Use With (Application) Wiping liquids or cleaning delicate glass, instruments, parts, and surfaces.
For Use With (Equipment) Suitable for ISO Class 4 or higher laboratory environments.
Packaging Quantity 100 Sheets x 24 Dispenser Boxes
Ply 2-ply