Learn More
DWK Life Sciences Kimble™ Tooled Neck N-51A Borosilicate Glass Vials with Unattached Polyethylene Closure
Product Code. 12380170
Click to view available options
Volume (Metric):
4 mL
12 mL
Height (Metric):
45 mm
50 mm
Diameter (Metric) Outer:
15 mm
21 mm
Volume (English):
1 dr.
3.2 dr.
Unit Size:
Pack of 432
Pack of 72
Pack of 864
Product Code. 12380170
Brand: DWK Life Sciences 60975L3
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Tooled neck OPTICLEAR™ vials are excellent for packaging drug products. Available in a range of capacities (4mL, 12mL, 20mL) in shelf packs of 72 .
  • Necks are tooled for an accurate fit with polyethylene closures
  • Vials and closures are supplied in both the pack and case quantities
  • Supplied with closed-bottom, two-piece closures
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
  • Options:
    • Capacity 4mL / 1 dram: Height 45mm, OD 15mm, Case of 864
    • Capacity 12mL / 3 drams: Height 50mm, OD 21mm, Case of 432
    • Capacity 20mL / 5 drams: Height 55mm, OD 27mm, Case of 216
    TRUSTED_SUSTAINABILITY
    Specifications
    Classification B
    Color Clear
    Diameter (Metric) Outer 21 mm
    Material Borosilicate Glass
    Volume (Metric) 12 mL
    Volume (English) 3.2 dr.
    Product Line Kimble
    Volume (Metric) Total 12 mL
    Closure Material Polyethylene
    Closure Type Open Bottom Stopper
    Height (Metric) 50 mm
    Quantity 72 Pk.
    Certifications/Compliance ASTM E438, USP Type I, Type I, Class B
    Type Vial