Learn More
DWK Life Sciences Kimble™ Solid Borosilicate Glass Beads for Distillation Columns
Product Code. p-89700
Change view
Click to view available options
Diameter (Metric):
5 mm
6 mm
Unit Size:
1lb
Pack of 3000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
15105205 5 mm Pack of 3000
14851192 6 mm 1lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15105205 Supplier DWK Life Sciences Supplier No. 135005
Resist attack by liquids or vapors
May be used as mixing beads, boiling stones, or packing
Order Info
Packed in 1 lb. boxes
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass
Diameter (Metric) 5 mm