Learn More
DWK Life Sciences Kimble™ Screw Caps for Disposable Glass Tubes
Description
Manufactured from polypropylene. Kimble Chase™ Screw Caps for Disposable Glass Tubes are autoclavable.
- Polypropylene
- Autoclavable
Compliance
Designed from ASTM™ E 982
Specifications
Specifications
| Certifications/Compliance | ASTM E982 |
| Closure Size | 13-415 |
| Product Type | Screw Cap |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Color | Natural |
| For Use With (Equipment) | Disposable Glass Tubes |