Learn More
DWK Life Sciences Kimble™ Kontes™ ULTRA-WARE™ Heavy-Wall Filter Flasks
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
278.46€ - 479.40€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11752754
|
DWK Life Sciences
953760-2002 |
2000 mL |
451.00€
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11702794
|
DWK Life Sciences
953760-4002 |
4 L |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
12851531
|
DWK Life Sciences
953760-1002 |
1000 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
Heavy-wall borosilicate glass provides mechanical strength for vacuum filtration
Kimble Chase KimCote™ plastic-coated glassware provides an added measure of safety in the event of breakage
- KimCote™ plastic safety coating
- ≠2 side arm for connection to 0..25 in. (6mm) I.D. flexible tubing
- 125mL flask: With No. 5 stopper joint
- 1, 2 and 4L flasks: With No. 8 stopper joint
Specifications
Borosilicate Glass |