Learn More
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™ Microtubes
Product Code. p-4883110
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
Material:
Polypropylene
RNase-, DNase-, pyrogen-free
Unit Size:
Pack of 100
Pack of 500
Product Code. 14824171
Brand: DWK Life Sciences 7495101501
For use with Pellet Pestle
  • Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
  • Individually wrapped
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Color Blue
Capacity (English) 0.05 oz.
Capacity (Metric) 1.5 mL
Material Polypropylene
Product Type Microtube
Dnase Free Yes
Capacity 0.05 oz., 1.5mL