Learn More
DWK Life Sciences Kimble™ Kontes™ Incubation Flasks
Product Code. 10208294
Change view
Click to view available options
Capacity (Metric):
10 mL
Closure Material:
Rubber
Material:
Borosilicate Glass
Includes:
Sidearm
w/o Sidearm
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Material Material Includes unitSize
10208294 10 mL Rubber Borosilicate Glass w/o Sidearm Pack of 12
11843173 10 mL Rubber Borosilicate Glass Sidearm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10208294 Supplier DWK Life Sciences Supplier No. 8823000010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For metabolic studies of biological materials
Ideal for measuring C14O2 yields following incubation, in vitro incubations, insulin bioassays, persulfate oxidations, and distillation of any volatile acid or base. Also suitable for use with phosphate or bicarbonate buffers when acid or alkali is introduced at the end of a reaction.
  • Available with or without sidearm
  • Flask with sidearm enables syringe needle entry without disturbing established conditions
Order Info
Stoppers and center well sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Closure Material Rubber
Capacity (Metric) 10 mL
Material Borosilicate Glass
For Use With (Application) For measuring C14O2 yields following incubation, in-vitro incubations, Insulin bioassays, Persulfate oxidations and distillation of any volatile Acid or base
Height (English) 2 in.
Height (Metric) 5 cm
Includes w/o Sidearm
Width (English) 1.2 in.
Width (Metric) 3.1 cm
Closure Type Stopper