Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders
Product Code. 11854005
Change view
Click to view available options
Capacity:
0.03 oz., 1mL
0.1 oz., 3mL
0.16 oz., 5mL
Includes:
Ground Glass pestle and tube
PTFE pestle and Glass tube
Size:
20
21
22
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Includes Size unitSize
11854005 0.03 oz., 1mL Ground Glass pestle and tube 20 Each
10271645 0.16 oz., 5mL PTFE pestle and Glass tube 22 Each
10545115 0.1 oz., 3mL Ground Glass pestle and tube 21 Each
10398022 0.03 oz., 1mL PTFE pestle and Glass tube 20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11854005 Supplier DWK Life Sciences Supplier No. 8854500020
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
Combined conical and cylindrical surfaces improve grinding efficiency
  • Can be motor driven
  • Choose from all-ground glass style or smooth glass tube with PTFE pestle threaded to stainless-steel shaft
  • Interchangeable tubes and pestles have 0.004 to 0.006 in. (0.10 to 0.15mm) clearance
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.03 oz.
Clearance (English) 0.004 to 0.006 in.
Clearance (Metric) 0.1 to 0.15mm
Diameter (Metric) Shaft 6mm
Diameter (English) Shaft 0.23 in.
Capacity (Metric) 1 mL
For Use With (Application) Ideal for connective tissue such as muscle, heart and lung
Includes Ground Glass pestle and tube
Length (English) Shaft 6.1 in.
Length (English) Tube 3.14 in.
Length (Metric) Shaft 155mm
Length (Metric) Tube 80mm
Tube Diameter (English) Outside 0.51 in.
Tube Diameter (Metric) Outside 13mm
Capacity 0.03 oz., 1mL
Size 20