Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders
Product Code. 10271645 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1 mL
3 mL
5 mL
Includes:
Ground Glass pestle and tube
PTFE pestle and Glass tube
Size:
20
21
22
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Includes Size Unit Size
10271645 5 mL PTFE pestle and Glass tube 22 Each
11854005 1 mL Ground Glass pestle and tube 20 Each
10545115 3 mL Ground Glass pestle and tube 21 Each
10398022 1 mL PTFE pestle and Glass tube 20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10271645 Supplier DWK Life Sciences Supplier No. 8854800022
  • ONLINE EXCLUSIVE PRICE
    205.15€
    140.33€ / Pack of 1
    Save 64.82€
    31% Off
Estimated Shipment: 22-01-2027
Combined conical and cylindrical surfaces improve grinding efficiency
  • Can be motor driven
  • Choose from all-ground glass style or smooth glass tube with PTFE pestle threaded to stainless-steel shaft
  • Interchangeable tubes and pestles have 0.004 to 0.006 in. (0.10 to 0.15mm) clearance
Specifications
Capacity (English) 0.16 oz.
Clearance (English) 0.004 to 0.006 in.
Clearance (Metric) 0.1 to 0.15mm
Diameter (Metric) Shaft 6mm
Diameter (English) Shaft 0.23 in.
Capacity (Metric) 5 mL
For Use With (Application) Ideal for connective tissue such as muscle, heart and lung
Includes PTFE pestle and Glass tube
Length (English) Shaft 8.97 in.
Length (English) Tube 5.9 in.
Length (Metric) Shaft 228mm
Length (Metric) Tube 150mm
Tube Diameter (English) Outside 0.7 in.
Tube Diameter (Metric) Outside 18mm
Size 22
Product Type Tissue Grinder