Learn More
DWK Life Sciences Kimble™ KIMAX™ Heavy-Duty Beakers
Click to view available options
Capacity (Metric):
250 mL
400 mL
1000 mL
2000 mL
Packungsgröße:
Pack of 12
Pack of 4
Pack of 6
Beschreibung
Heavy construction prolongs life
- With pouring spout
- With marking spot
- Have white graduations for approximating volume
- Accurate to ±5%
- Thick, uniform walls with extra wall thickness in tooled top rim
Compliance
Designed from ASTM™ E 960 Type II requirements.
Spezifikation
Spezifikation
| Accuracy | ±5% |
| Capacity (English) | 13.52 oz. |
| Diameter (English) Outer | 3.03 in. |
| Diameter (Metric) Outer | 7.7 cm |
| Capacity (Metric) | 400 mL |
| Material | Borosilicate Glass |
| For Use With (Application) | Offer Superior Mechanical Strength and Durability, Provides Improved Safety When Used Under Extreme Conditions Such as Mechanized Washing and Rough Handling |
| Graduation Range | 25 to 325 mL |
| Height (English) | 4.48 in. |
| Height (Metric) | 11.4 cm |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur