Learn More
DWK Life Sciences KIMBLE™ Glass Screw-Thread Sample Vials with PTFE Closure
Product Code. 15205547 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
4.0 mL
8.0 mL
Volume (English):
1 dr.
2.16 dr.
Unit Size:
Pack of 144
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Volume (English) Conditionnement
15205547 8.0 mL 2.16 dr. Pack of 144
12763067 4.0 mL 1 dr. Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15205547 Fournisseur DWK Life Sciences Code fournisseur 60940A8
  • ONLINE EXCLUSIVE PRICE
    159.00€
    148.61€ / Pack of 144
    Save 10.39€
    6% Off
Estimated Shipment: 28-12-2026
Ajouter au panier
Ajouter au panier
KIMBLE™ Glass Screw-Thread Sample Vials with PTFE Closure are clear borosilicate glass vials with a flat bottom for free-standing use. Made to USP Type I and ASTM E438 standards, each vial includes an attached white urea cap with PTFE-faced white rubber liner.
Cylindrical shape provides solid balance. Flat bottom facilitates free-standing capability.
  • Clear borosilicate glass sample vials with cylindrical shape and flat bottom for free-standing stability
  • Screw-thread GPI finish for secure, leak-resistant closure
  • Supplied with attached closed-top white urea cap with PTFE-faced white rubber liner
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • 60940A-4 complies with EPA 600 series methods
  • Packaged in corrugated trays with partitions
Specifications
Classification A
Color Clear
Diameter (Metric) Outer 17 mm
Material Borosilicate Glass
Pre-slit No
Sterility Non-sterile
Volume (Metric) 8.0 mL
Thread Style Screw
Volume (English) 2.16 dr.
Closure Material PTFE
Closure Type Screw Cap (Closed-top)
Quantity 144/Cs.
Marking Spot No
Autoclavable Autoclavable
Certifications/Compliance ASTM Type I, Class A
Height (Metric) 63 mm
Product Type Sample Vial with Screw Cap