Learn More
DWK Life Sciences Kimble™ KIMAX™ Conical-Bottom Glass Centrifuge Tubes: 15 mL Capacity
Product Code. 11850084
Change view
Click to view available options
Disposable:
Reusable
Single-use
Graduations:
0.1 mL
Permanent Red, 0.1 mL
Length (Metric):
118 mm
126 mm
130 mm
Unit Size:
Pack of 12
Pack of 125
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Disposable Graduations Length (Metric) unitSize
11850084 Reusable 0.1 mL 118 mm Pack of 12
11825801 Reusable Permanent Red, 0.1 mL 118 mm Pack of 12
11828542 Reusable - 118 mm Pack of 12
11888452 Reusable 0.1 mL 130 mm Pack of 12
11845115 Single-use - 126 mm Pack of 125
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11850084 Supplier DWK Life Sciences Supplier No. 4516515
Withstand centrifugation up to 2980 RCF
  • With conical shaped bottom
  • Tubes are made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class A or B glass requirements
TRUSTED_SUSTAINABILITY
Specifications
Calibration To contain
Certifications/Compliance USP, ASTM E 438
Capacity (Metric) 15 mL
Disposable Reusable
Material Borosilicate Glass
Tolerance 0 to 0.2mL in ±0.05mL, >0.2 to 1mL in ±0.1mL, >1 to 3mL in ±0.15mL, >3mL in ±0.2mL
Shape Conical
Length (Metric) 118 mm
Bottom Shape Conical
Closure Type Not Included
Writing Area Marking Spot
Diameter (Metric) 17 mm
Graduations 0.1 mL
Product Type Centrifuge Tube
Max. RCF 2980 x g