Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Bonded White PTFE Liners
Product Code. 11833043
Change view
Click to view available options
Color:
Black
Blue
White
Closure Size:
13-425
8-425
9-425
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Closure Size unitSize
11833043 White 8-425 Pack of 1000
11823043 Blue 9-425 Pack of 1000
11813043 Black 13-425 Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11833043 Supplier DWK Life Sciences Supplier No. 73814WH8425
Intended for single-use injection
  • Manufactured using the highest quality raw materials
  • Designed for an ideal fit with Kimble Chase autosampler vials
  • Pre-assembled caps and septa minimizes contamination due to handling
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) PTFE
Color White
Material Polypropylene
Closure Size 8-425