Learn More
DWK Life Sciences Kimble™ Kontes™ Weighing/Transfer Funnels
Product Code. 15376657
Change view
Click to view available options
Height (Metric):
13 mm
Diameter (Metric)Top:
60 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Diameter (Metric)Top Unit Size
15376657 13 mm 60 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15376657 Supplier DWK Life Sciences Supplier No. 7476000013
  • ONLINE EXCLUSIVE PRICE
    45.99€
    28.04€ / Pack of 1
    Save 17.95€
    39% Off
 Restricted Item
Estimated Shipment: 30-04-2027
For easy transfer of liquids or powders
  • Flat bottom
Specifications
Diameter (Metric)Top 60 mm
Material Borosilicate Glass
Height (Metric) 13 mm
For Use With (Application) Used for easy transfer of liquids or powders
Product Type Transfer/Weighing Funnel