Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Tube For All-Glass Dounce Tissue Grinders
Product Code. 11883722 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
2 mL
7 mL
15 mL
40 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11883722 2 mL Each
10432385 7 mL Each
11816271 15 mL Each
11864361 40 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11883722 Supplier DWK Life Sciences Supplier No. 8853030002
  • ONLINE EXCLUSIVE PRICE
    82.20€
    77.54€ / Pack of 1
    Save 4.66€
    5% Off
 Restricted Item
Estimated Shipment: 05-01-2027
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Capacity (English) 0.06 oz.
Capacity (Metric) 2 mL
Material Borosilicate Glass
Product Type Tissue Grinder Tube
For Use With (Equipment) Dounce all Glass tissue grinders
Quantity Case of 1