Learn More
DWK Life Sciences Kimble™ Kontes™ Pestles For Dounce All-Glass Tissue Grinders
Product Code. 10389444
Change view
Click to view available options
Capacity:
0.06 oz., 2mL
0.23 oz., 7mL
Product Type:
Large Clearance Pestle, A
Small Clearance Pestle, B
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Product Type unitSize
10389444 0.06 oz., 2mL Small Clearance Pestle, B Each
13161124 0.06 oz., 2mL Large Clearance Pestle, A Each
10581133 0.23 oz., 7mL Small Clearance Pestle, B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10389444 Supplier DWK Life Sciences Supplier No. 8853020002
All-glass component parts.
  • For use with Dounce (Kontes 885300 series) tissue grinders
  • B (Tight)
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 0.06 oz.
Capacity (Metric) 2 mL
Product Type Small Clearance Pestle, B
For Use With (Equipment) Dounce tissue grinder
Capacity 0.06 oz., 2mL