Learn More
DWK Life Sciences Kimble™ Kontes™ Microscale B - Connector
Product Code. 17760878
Change view
Click to view available options
Thread Size:
13-425
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
17760878 13-425 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17760878 Supplier DWK Life Sciences Supplier No. 7471850013
  • ONLINE EXCLUSIVE PRICE
    21.84€
    19.66€ / Pack of 1
    Save 2.18€
    9% Off
Estimated Shipment: 17-02-2027
Unique design adapts GPI threaded glass to a serrated PTFE hose connection or 0.25 in. -28 tubing connections
  • Fits hose I.D.: 0.25 in.
  • Internal Thread Size: 0.25 in.-28
  • GPI Thread Size: 13-425
Specifications
Material PTFE
Thread Size 13-425
Product Type Hose Connector