Learn More
DWK Life Sciences KIMBLE™ KONTES™ Flow-Watchman™ Water Flow Indicator
Product Code. 11882923 Shop All DWK Life Sciences Products
Change view
Click to view available options
Case Material:
Polycarbonate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Case Material Unit Size
11882923 Polycarbonate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11882923 Supplier DWK Life Sciences Supplier No. 6262500000
  • ONLINE EXCLUSIVE PRICE
    31.25€
    26.32€ / Pack of 1
    Save 4.93€
    15% Off
 Restricted Item
Estimated Shipment: 16-02-2027
The KIMBLE™ KONTES™ Flow-Watchman™ Water Flow Indicator inserts into water lines for an instant check of flow continuity and relative flowrate. A white nylon ball spins at speeds proportional to flow velocity, visible from 25 ft., supporting flowrates from 100 mL/min to 1 L/min.
Insert in water lines for an instant check of flow continuity and relative flowrate.
  • White nylon ball spins through circular course at speed that increases with velocity of water flow
  • Visible from 25 ft. (8 m)
  • Usable with flowrates from 100 mL/min to 1 L/min
  • Maximum pressure: 10 psig
  • Tapered inlet and outlet fit No. 1 hose connections
  • Clear polycarbonate construction
Order Info
Dimensions: 1/2L x 3W x 2 in.H (1 x 8 x 5cm).
Specifications
Type Flow Indicator
Airflow Rate 100 mL/min. to 1 L/min.
Case Material Polycarbonate
Float Material Nylon
Pressure Rating 10 psig
Dimensions (L x W x H) 0.5 x 3 x 2 in. (1 x 8 x 5 cm)
Flow Rate Range 100 to 1 LPM
Height (English) 2 in.
Height (Metric) 5 cm
Length (English) 0.5 in.
Length (Metric) 1 cm
Width (Metric) 8 cm
Width (English) 3 in.