Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn™ Economy Columns
Click to view available options
Length (Metric):
40 mm
100 mm
300 mm
500 mm
Column Capacity:
1.5 mL
4 mL
12 mL
19 mL
Quantity:
1
10
Unit Size:
Each
Pack of 10
Description
Use columns with ion-exchange, affinity, gel filtration, and hydrophobic interaction media.
- Durable borosilicate glass
- Precision-ground column ends form positive seals with endcaps so they don't leak or “creep” under pressure
- Threaded, easy to remove reservoir cap won't pop off under pressure
- Reservoir accepts accessory funnels and glass reservoirs
- Luer-Lock polypropylene inlet and outlet fittings
- Bed support has a 20μm polyethylene disc that retains most media for low-pressure aqueous chromatography
- Columns accept flow adapters for adjusting Flex-Column bed height (order adapters separately)
Flow adapters not available
Spécification
Spécification
| Endcapped | Yes |
| For Use With (Application) | Ion exchange, affinity, gel filtration and hydrophobic interaction media |
| Quantity | 10 |
| Temperature (Metric) Range | 0 to 50°C |
| Diameter (Metric) Inner | 7 mm |
| Type | Economy Chromatography Column |
| Max. Pressure | 14.5 psi |
| Pore Size | 20 μm |
| Column Capacity | 1.5 mL |
| Length (Metric) | 40 mm |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu