Learn More
DWK Life Sciences Kimble™ Kontes™ Extraction Manifold Replacement Components
Product Code. 10714017
Change view
Click to view available options
Type:
Glass Funnel (300 mL)
PTFE Gasket
SPE Glassware Set (47 mm)
Support Base
Unit Size:
Each
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10714017 SPE Glassware Set (47 mm) Each
11803143 PTFE Gasket Pack of 25
10432635 Glass Funnel (300 mL) Each
10440165 Support Base Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10714017 Supplier DWK Life Sciences Supplier No. 9711000047
  • ONLINE EXCLUSIVE PRICE
    769.65€
    460.84€ / Pack of 1
    Save 308.81€
    40% Off
 Restricted Item
Estimated Shipment: 16-11-2026
For extraction manifold.
  • For use with Kontes SPE Manifold Sets (971000-1047, -3047, or -6047)
  • Kontes SPE Glassware and Manifolds are specially designed as support systems for 47mm and 90mm dia. solid phase extraction disks to give you the highest possible recoveries of extracted analytes from your drinking water, surface water and waste water samples
Specifications
For Use With (Equipment) Solid Phase Extraction Manifold
Type SPE Glassware Set (47 mm)
Includes 47 mm Funnel, support base, support screen, PTFE gaskets and clamp
Material Glass
Length (Metric) 47 mm