Learn More
DWK Life Sciences Kimble™ Kontes™ Extraction Manifold Replacement Components
Product Code. 10440165
Click to view available options
Type:
Glass Funnel (300 mL)
PTFE Gasket
SPE Glassware Set (47 mm)
Support Base
Unit Size:
Each
Pack of 25
Product Code. 10440165
Brand: DWK Life Sciences 9711020047
Only null left
Add to basket
Add to basket
For extraction manifold.
  • For use with Kontes SPE Manifold Sets (971000-1047, -3047, or -6047)
  • Kontes SPE Glassware and Manifolds are specially designed as support systems for 47mm and 90mm dia. solid phase extraction disks to give you the highest possible recoveries of extracted analytes from your drinking water, surface water and waste water samples
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Solid Phase Extraction Manifold
Type Support Base
Size (English) Filter 1.85 in.
Size (Metric) Filter 4.7 cm
Length (Metric) 47 mm