Learn More
DWK Life Sciences Kimble™ Kontes™ Component Parts for 25mm Ultra-Ware™ Microfiltration Assembly
Product Code. 11818482
Change view
Click to view available options
Support Base; 25mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11818482 Support Base; 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11818482 Supplier DWK Life Sciences Supplier No. 9537020001
Component parts for 25mm Ultra-Ware ™microfiltration assembly
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ultra-ware microfiltration assembly with fritted glass support
Type Support Base
Diameter (Metric) Inside 25 mm