Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column Accessories
Product Code. 17790878
Change view
Click to view available options
Product Type:
Drying Tube/Chromatography Column
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
17790878 Drying Tube/Chromatography Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17790878 Supplier DWK Life Sciences Supplier No. 7473000050
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
For chromatography columns
  • 747000 Series Microflex Kits
  • Features versatile threaded chaste connections
  • Can be assembled rightside up or upside down, providing great versatility
  • GPI thread size: 13 to 425
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For chromatography columns
Thread Size 13-425
Product Type Drying Tube/Chromatography Column