Learn More
DWK Life Sciences Kimble™ Kontes™ Adsorption/Ion-Exchange Columns
Click to view available options
Length (Metric):
300 mm
400 mm
500 mm
600 mm
Diameter (Metric) Inner:
11 mm
19 mm
22 mm
25 mm
41 mm
Column Capacity:
20 mL
110 mL
250 mL
790 mL
Includes:
Fritted disc
Without fritted disc
Unit Size:
Each
Description
- Borosilicate glass
- Sealed-in coarse porosity (40 to 60μm) fritted disc supports packing in K420320 Series columns
- Use glass wool to support packing in K420300 Series columns (packing not included)
Specifications
Specifications
| For Use With (Application) | Many separations in adsorption and ion exchange chromatography |
| Diameter (Metric) Inner | 22 mm |
| Type | Adsorption/Ion Exchange Chromatography Column |
| Pore Size | 40 to 60 μm |
| Column Capacity | 110 mL |
| Length (Metric) | 300 mm |
| Material | Borosilicate Glass |
| Includes | Fritted disc |