Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Microfiltration Assembly Component Parts
Product Code. 11803123
Click to view available options
Type:
Clamp
Funnel
Support Base (47 mm)
Support Base (90 mm)
Unit Size:
Each
Product Code. 11803123
Brand: DWK Life Sciences 9537530000
Components for Ultra-ware™ microfiltration assembly
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Ultra-ware microfiltration assembly with PTFE-faced fritted glass support
Material Aluminum
Type Clamp