Learn More
DWK Life Sciences Kimble™ KIMAX™ Flasks with Quick-Release Hose Connector
Product Code. 11757989
Click to view available options
Capacity (Metric):
250 mL
2000 mL
4 L
Unit Size:
Each
Pack of 2
Product Code. 11757989
Brand: DWK Life Sciences 270704000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Allow safe attachment of 0.25 in. I.D. (6mm) tubing
  • Tubing remains permanently attached to hose barb and opposite end attaches to flask tubulation with positive threaded seal
  • Proper positioning of connector provides greater flask stability as tubing angled downward has less tendency to tilt flask
  • Graduated with approximate volume scale in durable white ceramic enamel
  • Tubing and stoppers sold separately
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 8.18 in.
Capacity (Metric) 4 L
Material Borosilicate Glass
Graduation Range 1000 to 4000 mL
Graduations 250 mL
Height (English) 12.12 in.
Height (Metric) 30.8 cm
Diameter (Metric) Outer 20.8 cm
Closure Size 12