Learn More
DWK Life Sciences™ Kimble™ KIMAX™ Distilling Apparatus Complete Assembly
Product Code. p-169137
Click to view available options
Capacity:
33.81 oz., 1000mL
Unit Size:
Each
Product Code. 12754037
Brand: DWK Life Sciences™ 215001000
Includes
  • 500 or 1000mL flat-bottom flask
  • Distillation head with a standard taper 19/38 joint stopper
  • Jacketed (200mm) Graham condenser with standard taper 19/38 top joint
Complete assembly
Use to distill small amounts of water or in APHA Method 4500-NH3 for determining nitrogen (ammonia) in purified drinking water, natural water and highly purified wastewaters (concentration, <20μg/L)
TRUSTED_SUSTAINABILITY
Specifications
Includes 500 or 1000mL flat-bottom flask, distillation head with a 19/38 joint stopper, jacketed graham condenser with 19/38 top joint
For Use With (Application) General purpose distillation and phenol distillation
Type Distillation Apparatus
Material Borosilicate Glass
Capacity 33.81 oz., 1000mL
Certifications/Compliance USP Type I and ASTM E438, Type I, Class A
Capacity (Metric) 1000mL
Standard Taper Stopper No. 27
Standard Taper Joint 19/38
Capacity (English) 33.81 oz.