Learn More
DWK Life Sciences Kimble™ Clear Screw Thread Sample Vials without Closures
Product Code. 11842983
Change view
Click to view available options
Volume (Metric):
1.8 mL
2 mL
4 mL
6 mL
8 mL
12 mL
16 mL
20 mL
24 mL
25 mL
40 mL
Diameter (Metric) Outer:
12 mm
15 mm
17 mm
19 mm
21 mm
23 mm
28 mm
Closure Size:
13-425
15-425
18-400
20-400
24-400
8-425
Unit Size:
Pack of 200
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Diameter (Metric) Outer Closure Size unitSize
11842983 2 mL 15 mm 13-425 Pack of 200
11822993 20 mL 28 mm 24-400 Pack of 200
11882983 6 mL 19 mm 15-425 Pack of 200
11832993 25 mL 28 mm 24-400 Pack of 200
11842993 40 mL 28 mm 24-400 Pack of 200
11862983 4 mL 17 mm 15-425 Pack of 200
11822983 1.8 mL 12 mm 8-425 Pack of 200
11852983 4 mL 15 mm 13-425 Pack of 200
11812993 24 mL 23 mm 20-400 Pack of 200
11892983 12 mL 19 mm 15-425 Pack of 200
11802993 16 mL 21 mm 18-400 Pack of 200
11832983 2 mL 12 mm 8-425 Pack of 200
11872983 8 mL 17 mm 15-425 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11842983 Supplier DWK Life Sciences Supplier No. 608121528
Packaged in corrugated trays with partitions
  • Manufactured from clear 33 expansion borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Diameter (Metric) Outer 15 mm
Material Borosilicate Glass
Volume (Metric) 2 mL
Thread Style Screw
Volume (English) 0.5 dr.
Closure Size 13-425
Certifications/Compliance ASTM Type I, Class A
Height (Metric) 28 mm