Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures
Product Code. 11873003
Change view
Click to view available options
Volume (English):
0.5 dr.
1 dr.
1.5 dr.
10 dr.
2 dr.
5 dr.
Closure Size:
13-425
15-425
24-400
8-425
Unit Size:
Pack of 200
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (English) Closure Size unitSize
11873003 0.5 dr. 13-425 Pack of 200
11863003 0.5 dr. 8-425 Pack of 200
11803013 2 dr. 15-425 Pack of 200
11813013 1.5 dr. 15-425 Pack of 200
11883003 1 dr. 13-425 Pack of 200
11833013 10 dr. 24-400 Pack of 200
11823013 5 dr. 24-400 Pack of 200
11893003 1 dr. 15-425 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11873003 Supplier DWK Life Sciences Supplier No. 609121528
Manufactured from amber 33 expansion borosilicate glass
  • Amber vials offer protection against harmful light and UV rays for light sensitive products.
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Diameter (Metric) Outer 15 mm
Material Borosilicate Glass
Volume (Metric) 2 mL
Thread Style Screw
Volume (English) 0.5 dr.
Closure Type Screw Cap
Closure Size 13-425
Certifications/Compliance USP Type I
Height (Metric) 28 mm