Learn More
Kimberly-Clark Professional™ Kimtech™ M3 Sterile Face Masks
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
16.80€
Specifications
Product Type | Cleanroom Face Mask |
---|---|
Color | White |
Material | SMS |
Size | Standard |
Sterility | Sterile |
Product Code | Brand | Fastening Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Fastening Type | Includes | Price | Quantity & Availability | ||||
11847531
|
Kimberly-Clark Professional™
62494 |
Ties | With 9 in. soft ties |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
- Gamma-sterilized to 10-6
- White with soft ties or earloops
- Outer layer of apertured polyethylene film to provide protection from contamination
- Polyester binding and polyester filter media provide bacteria filtration efficiency of 91.9% and particle filtration efficiency of 93.1%
Specifications
Cleanroom Face Mask | |
SMS | |
Sterile | |
Class 1 (ISO 3) | |
89.3% (PFE), 94.1% (BFE) | |
0.1 μm (PFE), 3 μm (BFE) |
White | |
Standard | |
Pleated | |
Yes | |
Yes | |
Kimtech™ M3 Sterile |