Learn More
Kimberly-Clark Professional™ KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves
Product Code. 15988156
Change view
Click to view available options
:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15988156 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15988156 Supplier Kimberly-Clark Professional™ Supplier No. 38718
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Low-lint gloves are ideal for detecting imperfections and defects when conducting critical quality control inspections. Ambidextrous gloves are silicone-free for use in a wide variety of applications including film handling, aerospace, and automotive assembly.
  • Ambidextrous design allows gloves to be replaced one at a time, rather than in pairs
  • High degree of sensitivity and dexterity for inspecting for defects and imperfections
  • Designed for critical quality control inspections
  • Seamless knit nylon shell reduces skin irritation
  • Low lint for cleanroom applications
  • Silicone-free
TRUSTED_SUSTAINABILITY
Specifications
Construction Seamless Knit
Coating Material Uncoated
Color White
Hand Ambidextrous
Material Nylon
Product Type Inspection Gloves
Size Medium
For Use With (Application) Automotive Assembly, Aerospace Engineering, and Quality Control Inspections
Glove Cuff Style Knit
Series KleenGuard™ G35