Learn More
DWK Life Sciences Kimble™ KIMAX™ Tall-Form Beakers, with spout
Product Code. 11730704
Click to view available options
Capacity (Metric):
200 mL
300 mL
Unit Size:
Pack of 12
Product Code. 11730704
Brand: DWK Life Sciences 14030200
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
With spout for easy pouring.
  • Matte-finish marking spot
  • Graduated to indicate approximate volumes
  • Berzelius style with flared top, reinforcing bead
Compliance
Designed from ASTM™ E 960 Type III (with spout) and Type IV (without spout) requirements
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 6.76 oz.
Diameter (English) Outer 2.24 in.
Diameter (Metric) Outer 5.7 cm
Capacity (Metric) 200 mL
Material Borosilicate Glass
For Use With (Application) For Performing Titrations
Graduation Range 25 to 150 mL
Height (English) 4.09 in.
Height (Metric) 10.4 cm
Includes Spout
Increments 25 mL
Label Type Matte Finish Marking
Quantity 12 Pk.
Type Beaker
Disposable Reusable
Graduated Yes
Certifications/Compliance ASTM E960, Type III, USP Type I, ASTM E438, Type I, Class A
Scale Dual White Scale
Style Tall Flared Top