Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Tubes with PTFE-Faced Rubber-Lined Screw Cap
Product Code. 11838452
Click to view available options
Diameter (Metric):
13 mm
16 mm
20 mm
25 mm
Length (Metric):
100 mm
125 mm
150 mm
200 mm
Capacity (Metric):
8 mL
12 mL
16 mL
20 mL
25 mL
30 mL
50 mL
70 mL
Thread Size:
13-415
15-415
18-415
24-410
Unit Size:
Pack of 144
Pack of 192
Pack of 288
Product Code. 11838452
Brand: DWK Life Sciences 45066A20150
Only null left
Add to basket
Add to basket
Black phenolic cap with PTFE-faced rubber liner
  • White marking spot
  • Black phenolic cap
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM E982, Type VI, Class B
Closure Color Black
Closure Material Phenolic
Capacity (Metric) 30 mL
Length (Metric) 150 mm
Material Borosilicate Glass
Label Type Marking Spot
Liner Material PTFE, Rubber
Thread Size 18-415
Diameter (Metric) 20 mm
Closure Type Screw Cap
Product Type Reusable Test Tube