Learn More
DWK Life Sciences Kimble™ KIMAX™ Griffin Beakers
Product Code. 11716333
Click to view available options
Capacity (Metric):
10 mL
20 mL
30 mL
100 mL
150 mL
250 mL
1000 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 11716333
Brand: DWK Life Sciences 140001000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
With thick, beaded, slightly flared top
  • With pouring spout
  • Matte-finish enameled marking spot
  • 20 to 4000mL sizes have white scales for approximate volume
  • Accurate to ±5%
  • Uniform, sturdy walls
Compliance
Designed from ASTM™ E 960 Type I requirements
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±5%
Capacity (English) 33.81 oz.
Capacity (Metric) 1000 mL
Material Borosilicate Glass
For Use With (Application) For Measuring and Mixing Liquids
Graduation Range 50 to 1000 mL
Includes Spout
Increments 50 mL
Label Type Matte Finish Enamel Marking
Quantity 6 Pk.
Type Beaker
Graduated Yes
Certifications/Compliance ASTM E960, Type I
Scale Dual White Scale
Style Griffin Beaded Top