Learn More
DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Serological Pipets with Tooled Opening
Product Code. 11858472
Change view
Click to view available options
Volume (Metric):
0.2 mL
1 mL
5 mL
10 mL
25 mL
Graduations:
0.01 mL
0.1 mL
Color:
Black
Blue
Orange
Red
White
Unit Size:
Pack of 12
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Color unitSize
11858472 0.2 mL 0.01 mL Black Pack of 6
11878532 1 mL 0.1 mL Red Pack of 12
11888472 5 mL 0.1 mL Blue Pack of 12
11878472 25 mL 0.1 mL White Pack of 12
11848472 10 mL 0.1 mL Orange Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11858472 Supplier DWK Life Sciences Supplier No. 37034A210
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Order Info
Plugs not included.
Compliance
Calibrated To Deliver/Blow Out. Designed from ASTM™ E 1044.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM E1044
Material Borosilicate Glass
Graduated Yes
Graduations 0.01 mL
Length (English) 11.81 in.
Tolerance ±0.008 mL
Color Black
Volume (Metric) 0.2 mL
Length (Metric) 295 mm
Type Serological Pipet