Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Product Code. 11758283
Change view
Click to view available options
Color:
Natural
Yellow
For Use With (Equipment):
13 mm Plain-end Culture Tubes
16 mm Plain-end Culture Tubes
18 mm Plain-end Culture Tubes
20 mm Plain-end Culture Tubes
25 mm Plain-end Culture Tubes
38 mm Plain-end Culture Tubes
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) unitSize
11758283 Natural 13 mm Plain-end Culture Tubes Pack of 500
10799335 Natural 20 mm Plain-end Culture Tubes Pack of 500
11778283 Natural 18 mm Plain-end Culture Tubes Pack of 500
10399924 Natural 16 mm Plain-end Culture Tubes Pack of 1000
11760734 Natural 25 mm Plain-end Culture Tubes Pack of 250
10349594 Natural 38 mm Plain-end Culture Tubes Pack of 250
10638205 Yellow 25 mm Plain-end Culture Tubes Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11758283 Supplier DWK Life Sciences Supplier No. 7366013
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Push Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
Sterility Non-sterile
For Use With (Equipment) 13 mm Plain-end Culture Tubes
Disposable Reusable