Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Product Code. 10799335
Change view
Click to view available options
Color:
Natural
Yellow
For Use With (Equipment):
13 mm Plain-end Culture Tubes
16 mm Plain-end Culture Tubes
18 mm Plain-end Culture Tubes
20 mm Plain-end Culture Tubes
25 mm Plain-end Culture Tubes
38 mm Plain-end Culture Tubes
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color For Use With (Equipment) Unit Size
10799335 Natural 20 mm Plain-end Culture Tubes Pack of 500
11778283 Natural 18 mm Plain-end Culture Tubes Pack of 500
11758283 Natural 13 mm Plain-end Culture Tubes Pack of 500
10399924 Natural 16 mm Plain-end Culture Tubes Pack of 1000
11760734 Natural 25 mm Plain-end Culture Tubes Pack of 250
10349594 Natural 38 mm Plain-end Culture Tubes Pack of 250
10638205 Yellow 25 mm Plain-end Culture Tubes Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10799335 Supplier DWK Life Sciences Supplier No. 7366020
  • ONLINE EXCLUSIVE PRICE
    132.00€
    117.92€ / Pack of 500
    Save 14.08€
    10% Off
 Restricted Item
Estimated Shipment: 19-02-2027
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
Specifications
Product Type Push Cap
Autoclavable Autoclavable
Material Polypropylene
Color Natural
Sterility Non-sterile
For Use With (Equipment) 20 mm Plain-end Culture Tubes
Disposable Reusable