missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX KILR™ MCF7 Cell Pool

Product Code. 17384849
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17384849 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17384849 Supplier DiscoverX Supplier No. 971014P028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The KILR™ MCF7 Cell Pool allows for assay of effector-induced cell death by a variety of immune mechanisms. This cell pool ships as two vials of cryopreserved cells.

The KILR MCF7 cell pool is a stable, engineered target cell pool which more closely reflects the heterogeneous native protein expression of tumor environments, used to measure direct cell death mediated by immunooncology treatments (e.g. checkpoint inhibitors, checkpoint agonists, bispecifics, CAR-T, and more) via Antibody Dependent Cell-mediated Cytotoxicity (ADCC), Complement Dependent Cytotoxicity (CDC), and Antibody Dependent Cell Phagocytosis (ADCP). This product uses EFC technology. The cell pool is a heterogeneous population of MCF7 cells that overexpress a PL-tagged housekeeping gene. Once the cells have been lysed by effector cells, PL is released to the media, where EA contained in the detection kit produces an EFC signal proportional to the level of PL released. This allows for calculation of percent lysis of the KILR cell pool. This stable cell pool is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Cytoxicity
Cell Line MCF7
For Use With (Application) Cell Pools
Quantity 2 Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.