Learn More
Honeywell Safety Products Kernmantel edge shock absorbing lanyard 1,5m + 1QT +1GO65 ET
Brand:  Honeywell Safety Products 1032379
Kernmantel edge shock absorbing lanyard 1,5m + 1QT +1GO65 ET
Product Code. 16081522
  • ONLINE EXCLUSIVE PRICE
    169.05€
    88.81€ / Each
    Save 80.24€
    47% Off
Only null left
Estimated Shipment: 19-03-2026