Learn More
DWK Life Sciences™ KECK™ Clip, for conical joints, from POM
Product Code. 10643353 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 40
NS 45
Color:
Amber
Blue
Green
Orange
Red
Violet
Yellow
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Color unitSize
10643353 NS 14 Yellow Pack of 10
10404672 NS 19 Blue Pack of 10
10011242 NS 24 Green Pack of 10
10571803 NS 19 Green Pack of 10
10264632 NS 12 Violet Pack of 10
10749474 NS 34 Orange Pack of 10
10254682 NS 45 Amber Pack of 10
10592183 NS 40 Yellow Pack of 10
11699808 NS 29 Red Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10643353 Supplier DWK Life Sciences™ Supplier No. 1090978
Only null left
Add to basket
Add to basket
The KECK™ clamp is used to secure a standard ground joint connection.
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • For conical joints
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
  • Increased safety in the laboratory as the glass surface is not scratched
  • Marking of the individual sizes for better differentiation
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
Material POM
For Use With (Equipment) Secure Glass-to-Glass Joints
For Use With (Application) Secure glass-to-glass joints
Size NS 14
Color Yellow