Learn More
DWK Life Sciences™ KECK™ Clip Assortment, for conical joints, from stainless steel (1.4310, blank)
Product Code. 10662183 Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 10
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 7
Unit Size:
Pack of 10
Product Code. 10662183
Brand: DWK Life Sciences™ 290301104
The KECK™ clamp is used to secure a standard ground joint connection.
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
Material Stainless Steel (1.4310, Blank)
For Use With (Equipment) Secure Glass-to-Glass Joints
For Use With (Application) Secure glass-to-glass joints
Size NS 34