Learn More
DWK Life Sciences™ KECK™ Clip Assortment, for conical joints, from POM
Product Code. 10233501 Shop All DWK Life Sciences Products
Click to view available options
For Use With (Equipment):
Secure Glass-to-Glass Joints
Unit Size:
Each
Product Code. 10233501
Brand: DWK Life Sciences™ 290310006
Only null left
Add to basket
Add to basket
The KECK™ clamp is used to secure a standard ground joint connection.
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • For conical joints
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
  • Increased safety in the laboratory as the glass surface is not scratched
  • Marking of the individual sizes for better differentiation
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Material POM
Includes 2 x KC 14, 2 x KC 19, 1 x KC 29, 1 x KC 10, 1 x KC 24, 1 x KC 34, 1 x KC 45
For Use With (Equipment) Secure Glass-to-Glass Joints
For Use With (Application) Secure glass-to-glass joints