Learn More
DWK Life Sciences™ KECK™ Adapter KA, complete, from glass thread to hose connection plastic
Product Code. 15854698 Shop All DWK Life Sciences Products
Click to view available options
Size:
GL 14, diameter 4.5 mm
GL 14, diameter 9.0 mm
RD 14, diameter 4.5 mm
RD 14, diameter 9.0 mm
Unit Size:
Each
Pack of 10
Product Code. 15854698
Brand: DWK Life Sciences™ 286345208
KECK™ adapters protect glass threads from damage.
KECK™ adapters for glass threads are available for sizes GL 14 and RD 14. They protect the thread from damage and ensure a fast, secure and tight connection. The threaded cap is applied directly to the external thread of the connection piece. The hose part can be quickly connected to the threaded cap with a 1/4 turn and loosened again.
  • Screw the threaded cap directly onto the external thread of the connection piece
  • Quick connection and disconnection of the hose part to the threaded cap with only 1/4 turn
  • They protect the thread from damage and ensure a fast, secure and tight connection.
  • Available for glass threads sizes GL 14 and RD 14
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
For Use With (Equipment) Connection of a Hose Piece with a Threaded Cap
For Use With (Application) Connection of a hose piece with a threaded cap
Size RD 14, diameter 4.5 mm