Learn More
KD Scientific Borosilicate Autoclavable Glass Syringes
Brand:  KD Scientific 780880
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11941017
  • 655.00€ / Each
Estimated Shipment: 02-07-2024
Specifications
Autoclavable
5 mL
200 mL
Borosilicate Glass
1/Pk.