Learn More
Fisherbrand™ KCR Quick Coupling Kit
Product Code. 11904169 Shop All Fisherbrand Products
Product Code. 11904169
Only null left
Add to basket
Add to basket
For fast connection and disconnection by simple snap-fastening. Fisherbrand™ KCR Quick Coupling Kits are designed for use with water faucets.
  • Comes with 15/21 tip and 20/37 adapter
  • Facilitates the water filtration and purification process
Specifications
Color Indicator No
For Use With (Application) For water purification
For Use With (Equipment) SI Wall Brackets
Includes 1-spigot, 1-fitting, 1 bi-conical quick coupling connection, 2 seals
Model KCR
Pressure 4 bar
Quantity 1 Kit
Type KCR Quick Coupling Kit