missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KCNF1 (Human) Recombinant Protein
Human KCNF1 full-length ORF ( AAH26110, 1 a.a. - 494 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003754-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDGSGERSLPEPGSQSSAASDDIEIVVNVGGVRQVLYGDLLSQYPETRLAELINCLAGGYDTIFSLCDDYDPGKREFYFDRDPDAFKCVIEVYYFGEVHMKKGICPICFKNEMDFWKVDLKFLDDCCKSHLSEKREELEEIARRVQLILDDLGVDAAEGRWRRCQKCVWKFLEKPESSCPARVVAVLSFLLILVSSVVMCMGTIPELQVLDAEGNRVEHPTLENVETACIGWFTLEYLLRLFSSPNKLHFALSFMNIVDVLAILPFYVSLTLTHLGARMMELTNVQQAVQALRIMRIARIFKLARHSSGLQTLTYALKRSFKELGLLLMYLAVGIFVFSALGYTMEQSHPETLFKSIPQSFWWAIITMTTVGYGDIYPKTTLGKLNAAISFLCGVIAIALPIHPIINNFVRYYNKQRVLETAAKHELELMELNSSSGGEGKTGGSRSDLDNLPPEPAGKEAPSCSSRLKLSHSDTFIPLLTEEKHHRTRLQSCK
Specifications
AAH26110 | |
potassium voltage-gated channel, subfamily F, member 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KCNF1 | |
GST |
3754 | |
Wheat germ expression system | |
25 μg | |
IK8, KCNF, KV5.1, MGC33316, kH1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |