Learn More
SI Analytics™ Karl-Fischer™ Titration Vessel
Shop All SI Analytics Products
Click to view available options
Unit Size:
Each
Description
- Constructed of glass
- Suitable for coulometry
- Includes one NS 19 opening and one NS 7.5 opening
- GL 18 thread size
Specifications
Specifications
| Type | Titration Vessel |
| For Use With (Application) | Coulometry |
| Includes | Openings |
| Material | Glass |
| Size | 1 x NS 19, 1 x NS 7.5 |