Learn More
Cole Parmer Kapton film 5mm, no adhesive 12inch by 12 inch
Brand:  Cole Parmer 08277-88
Kapton film 5mm, no adhesive 12inch by 12 inch
Product Code. 15302958
  • 62.55€ / Each
Only null left
Estimated Shipment: 04-02-2026