Learn More
Kallikrein 7 Goat anti-Human, Polyclonal, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 6 publications
Brand:  R&D Systems AF2624-SP
Encompass
Product Code. 15701574
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
Kallikrein 7 Polyclonal specifically detects Kallikrein 7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.